4-(3-Iodobenzyl)piperazine, N1-BOC protected 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: tert-Butyl 4-(3-iodobenzyl)piperazin-1-carboxylate

OR1425-1G
APOSOR1425-1GEA 174 EUR
APOSOR1425-1G
4-(3-Iodobenzyl)piperazine, N1-BOC protected 97%
4-(3-Iodobenzyl)piperazine, N1-BOC protected
Formel: C₁₆H₂₃IN₂O₂
Schmelzpunkt: 72…74 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06658978
CAS-Nummer: 850375-09-6
UN: 3259
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR