Tetrahydro-2H-pyran-4-carbonylchlorid 97%
Lieferant: Apollo Scientific
|
Gefahr
|
OR14254-5G
OR14254-10G
OR14254-1G
APOSOR14254-5GEA
0
EUR
APOSOR14254-5G
APOSOR14254-10G
APOSOR14254-1G
Tetrahydro-2H-pyran-4-carbonylchlorid 97%
Tetrahydro-2H-pyran-4-carbonylchlorid
Formel:
C₆H₉ClO₂ Siedepunkt: 50…52 °C/1mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD06200863 CAS-Nummer: 40191-32-0 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...