Ethyl-5-bromo-1-benzofuran-2-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl (5-bromobenzofuran)-2-carboxylate, Ethyl 5-bromobenzo[b]furan-2-carboxylate
OR14257-1G
APOSOR14257-1GEA
23.2
EUR
APOSOR14257-1G
Ethyl-5-bromo-1-benzofuran-2-carboxylate
Ethyl-5-bromo-1-benzofuran-2-carboxylate
Formel:
C₁₁H₉BrO₃ Molecular Weight: 269.1 g/mol Schmelzpunkt: 63…66 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02667598 CAS-Nummer: 84102-69-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...