[3-(2-Methyl-1,3-thiazol-4-yl)phenyl]methanol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3-(2-Methylthiazol-4-yl)phenyl)methanol, 4-[3-(Hydroxymethyl)phenyl]-2-methyl-1,3-thiazole, 3-(2-Methyl-1,3-thiazol-4-yl)benzyl alcohol
OR1426-100MG
APOSOR1426-100MGEA
34.8
EUR
APOSOR1426-100MG
[3-(2-Methyl-1,3-thiazol-4-yl)phenyl]methanol 97%
[3-(2-Methyl-1,3-thiazol-4-yl)phenyl]methanol
Formel:
C₁₁H₁₁NOS Molecular Weight: 205,276 g/mol Schmelzpunkt: 77,5…79.5 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02682066 CAS-Nummer: 850375-06-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...