5-Bromo-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)-1H-indole
Lieferant: Apollo Scientific
|
Achtung
|
OR14268-1G
APOSOR14268-1GEA
174
EUR
APOSOR14268-1G
5-Bromo-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)-1H-indole
5-Bromo-3-(1,4-dioxaspiro[4.5]dec-7-en-8-yl)-1H-indole
Formel:
C₁₆H₁₆BrNO₂ Molecular Weight: 334,22 g/mol Schmelzpunkt: 179…181 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06411613 CAS-Nummer: 282546-99-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...