2,3-Dihydro-1-benzofuran-2-ylmethanol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,3-Dihydro-2-(hydroxymethyl)benzo[b]furan, (2,3-Dihydrobenzofuran-2-yl)methanol
OR1427-5G
APOSOR1427-5GEA
318
EUR
APOSOR1427-5G
2,3-Dihydro-1-benzofuran-2-ylmethanol 97%
2,3-Dihydro-1-benzofuran-2-ylmethanol
Formel:
C₉H₁₀O₂ Molecular Weight: 150,18 g/mol Siedepunkt: 138 °C/9,5mmHg Dichte: 1,181 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD03086171 CAS-Nummer: 66158-96-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...