4-Chloro-N-(3-ethynylphenyl)benzenecarboxamide

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chloro-N-(3-ethynylphenyl)benzamide

OR14271-250MG
APOSOR14271-250MGEA 21.9 EUR
APOSOR14271-250MG
4-Chloro-N-(3-ethynylphenyl)benzenecarboxamide
4-Chloro-N-(3-ethynylphenyl)benzenecarboxamide
Formel: C₁₅H₁₀ClNO
Molecular Weight: 255,71 g/mol
Schmelzpunkt: 127…129 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00176067
CAS-Nummer: 883017-38-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR