2,4-Dichloro-1-(2-propynyloxy)benzene

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2,4-Dichloro-1-(prop-2-yn-1-yloxy)benzene

OR14273-250MG
APOSOR14273-250MGEA 27.7 EUR
APOSOR14273-250MG
2,4-Dichloro-1-(2-propynyloxy)benzene
2,4-Dichloro-1-(2-propynyloxy)benzene
Formel: C₉H₆Cl₂O
Molecular Weight: 201,05 g/mol
Schmelzpunkt: 47…49 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02311548
CAS-Nummer: 17061-90-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR