2-Hydroxy-2-(2-hydroxy-1-naphthyl)acetic acid

Lieferant: Apollo Scientific
Achtung

OR14276-250MG
APOSOR14276-250MGEA 63.8 EUR
APOSOR14276-250MG
2-Hydroxy-2-(2-hydroxy-1-naphthyl)acetic acid
2-Hydroxy-2-(2-hydroxy-1-naphthyl)acetic acid
Formel: C₁₂H₁₀O₄
Molecular Weight: 218,21 g/mol
Schmelzpunkt: 129…130 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06411618
CAS-Nummer: 624722-10-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR