4-[3-(Hydroxymethyl)benzyl]piperazine, N1-BOC protected

Lieferant: Apollo Scientific

Synonyme: tert-Butyl 4-[3-(hydroxymethyl)benzyl]piperazine-1-carboxylate, 3-{[4-(tert-Butoxycarbonyl)piperazin-1-yl]methyl}benzyl alcohol

OR1428-1G
APOSOR1428-1GEA 198 EUR
APOSOR1428-1G
4-[3-(Hydroxymethyl)benzyl]piperazine, N1-BOC protected
4-[3-(Hydroxymethyl)benzyl]piperazine, N1-BOC protected
Formel: C₁₇H₂₆N₂O₃
Molecular Weight: 306,4 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06797468
CAS-Nummer: 500013-39-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR