1,3-Oxazolo[4,5-b]pyridine-2-thiol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Thio-1,3-oxazolo[4,5-b]pyridine, Oxazolo[4,5-b]pyridine-2-thiol

OR14288-100MG
APOSOR14288-100MGEA 21.9 EUR
APOSOR14288-100MG
1,3-Oxazolo[4,5-b]pyridine-2-thiol
1,3-Oxazolo[4,5-b]pyridine-2-thiol
Formel: C₆H₄N₂OS
Schmelzpunkt: 241…243 °C
MDL: MFCD00973914
CAS-Nummer: 211949-57-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR