4-(4-(tert-Butoxycarbonyl)piperazin-1-yl)benzoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: Boc-piperazine-benzoic acid

OR14299-250MG
APOSOR14299-250MGEA 21.9 EUR
APOSOR14299-250MG
4-(4-(tert-Butoxycarbonyl)piperazin-1-yl)benzoic acid
4-(4-(tert-Butoxycarbonyl)piperazin-1-yl)benzoic acid
Formel: C₁₆H₂₂N₂O₄
Molecular Weight: 306,3569 g/mol
Schmelzpunkt: 50 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115067
CAS-Nummer: 162046-66-4
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR