5-Methyl-4-isoxazolcarbonsäure

Lieferant: Apollo Scientific
Achtung

OR14300-1G
APOSOR14300-1GEA 21.9 EUR
APOSOR14300-1G
5-Methyl-4-isoxazolcarbonsäure
5-Methylisoxazole-4-carboxylic acid
Formel: C₅H₅NO₃
Molecular Weight: 127,1 g/mol
Schmelzpunkt: 144…149 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00955655
CAS-Nummer: 42831-50-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR