[2-(1H-Pyrrol-1-yl)phenyl]methanol 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(1H-Pyrrol-1-yl)benzyl alcohol, (2-(1H-Pyrrol-1-yl)phenyl)methanol, 1-[2-(Hydroxymethyl)phenyl]-1H-pyrrole
OR1431-1G
APOSOR1431-1GEA
0
EUR
APOSOR1431-1G
[2-(1H-Pyrrol-1-yl)phenyl]methanol 97%
[2-(1H-Pyrrol-1-yl)phenyl]methanol
Formel:
C₁₁H₁₁NO Molecular Weight: 173,211 g/mol Siedepunkt: 104 °C (0,5 mm Hg) Schmelzpunkt: 42…44 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115123 CAS-Nummer: 61034-86-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...