Dimethylglyoxim 99%
Lieferant: Apollo Scientific
|
Gefahr
|
OR14325-100G
OR14325-2.5KG
OR14325-500G
APOSOR14325-100GEA
21.9
EUR
APOSOR14325-100G
APOSOR14325-2.5KG
APOSOR14325-500G
Dimethylglyoxim 99%
Dimethylglyoxim
Formel:
CH₃C(=NOH)C(=NOH)CH₃ Schmelzpunkt: 240…241 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002117 CAS-Nummer: 95-45-4 EINECS: 202-420-1 UN: 2926 ADR: 4.1+6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...