2-Amino-3-(methoxycarbonyl)benzoic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR14348-250MG
APOSOR14348-250MGEA
49.3
EUR
APOSOR14348-250MG
2-Amino-3-(methoxycarbonyl)benzoic acid 97%
2-Amino-3-(methoxycarbonyl)benzoic acid
Formel:
C₉H₉NO₄ Molecular Weight: 195,1721 g/mol Schmelzpunkt: 170…172 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06200897 CAS-Nummer: 253120-47-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...