Ethyl-3-aminothieno[2,3-b]pyridine-2-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 3-aminothieno[2,3-b]pyridine-2-carboxylate
OR14355-1G
APOSOR14355-1GEA
24.8
EUR
APOSOR14355-1G
Ethyl-3-aminothieno[2,3-b]pyridine-2-carboxylate
Ethyl-3-aminothieno[2,3-b]pyridine-2-carboxylate
Formel:
C₁₀H₁₀N₂O₂S Molecular Weight: 222,27 g/mol Schmelzpunkt: 183…185 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00173981 CAS-Nummer: 52505-46-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...