3',5-Dimethyl-3,5'-biisoxazole-4'-carboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Methyl-5-(5-methylisoxazol-3-yl)isoxazole-4-carboxylic acid

OR14356-1G OR14356-250MG
APOSOR14356-1GEA 325 EUR
APOSOR14356-1G APOSOR14356-250MG
3',5-Dimethyl-3,5'-biisoxazole-4'-carboxylic acid 97%
3',5-Dimethyl-3,5'-biisoxazole-4'-carboxylic acid
Formel: C₉H₈N₂O₄
Schmelzpunkt: 167 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01880302
CAS-Nummer: 849066-63-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR