(4-Aminophenyl)(4-cyclohexylpiperazino)methanone

Lieferant: Apollo Scientific
Achtung

OR14364-1G
APOSOR14364-1GEA 79.9 EUR
APOSOR14364-1G
(4-Aminophenyl)(4-cyclohexylpiperazino)methanone
(4-Aminophenyl)(4-cyclohexylpiperazino)methanone
Formel: C₁₇H₂₅N₃O
Molecular Weight: 287,41 g/mol
Schmelzpunkt: 134 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06659556
CAS-Nummer: 885949-70-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR