(4-Aminophenyl)[4-(2-pyrimidinyl)piperazino]methanone

Lieferant: Apollo Scientific
Achtung

OR14366-250MG
APOSOR14366-250MGEA 116 EUR
APOSOR14366-250MG
(4-Aminophenyl)[4-(2-pyrimidinyl)piperazino]methanone
(4-Aminophenyl)[4-(2-pyrimidinyl)piperazino]methanone
Formel: C₁₅H₁₇N₅O
Molecular Weight: 283,34 g/mol
Schmelzpunkt: 216 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659558
CAS-Nummer: 885949-72-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR