2-(1-Pyrrolyl)benzonitril

Lieferant: Apollo Scientific
Achtung

OR1437-1G OR1437-5G
APOSOR1437-1GEA 46.4 EUR
APOSOR1437-1G APOSOR1437-5G
2-(1-Pyrrolyl)benzonitril
2-(1-Pyrrolyl)benzonitril
Formel: C₁₁H₈N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00128308
CAS-Nummer: 33265-71-3
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR