2-(1-Pyrrolyl)benzonitril
Lieferant: Apollo Scientific
|
Achtung
|
OR1437-1G
OR1437-5G
APOSOR1437-1GEA
46.4
EUR
APOSOR1437-1G
APOSOR1437-5G
2-(1-Pyrrolyl)benzonitril
2-(1-Pyrrolyl)benzonitril
Formel:
C₁₁H₈N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00128308 CAS-Nummer: 33265-71-3 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...