2-(2-Chlorophenyl)-4-quinolinecarboxylic acid

Lieferant: Apollo Scientific
Achtung

OR14374-250MG
APOSOR14374-250MGEA 21.9 EUR
APOSOR14374-250MG
2-(2-Chlorophenyl)-4-quinolinecarboxylic acid
2-(2-Chlorophenyl)-4-quinolinecarboxylic acid
Formel: C₁₆H₁₀ClNO₂
Molecular Weight: 283,72 g/mol
Schmelzpunkt: 264…266 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00783457
CAS-Nummer: 20389-09-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR