4-(2-Hydroxyethyl)-2,5-dimethyl-1,2-dihydro-3H-pyrazol-3-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,2-Dihydro-2,5-dimethyl-4-(2-hydroxyethyl)-3H-pyrazol-3-one

OR14377-250MG
APOSOR14377-250MGEA 59.6 EUR
APOSOR14377-250MG
4-(2-Hydroxyethyl)-2,5-dimethyl-1,2-dihydro-3H-pyrazol-3-one
4-(2-Hydroxyethyl)-2,5-dimethyl-1,2-dihydro-3H-pyrazol-3-one
Formel: C₇H₁₂N₂O₂
Molecular Weight: 156,19 g/mol
Schmelzpunkt: 78…81 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03617955
CAS-Nummer: 258281-02-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR