Methyl-3-bromo-2,4-dioxo-4-phenylbutanoate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3-bromo-2,4-dioxo-4-phenylbutyrate

OR14388-1G
APOSOR14388-1GEA 132 EUR
APOSOR14388-1G
Methyl-3-bromo-2,4-dioxo-4-phenylbutanoate
Methyl-3-bromo-2,4-dioxo-4-phenylbutanoate
Formel: C₁₁H₉BrO₄
Molecular Weight: 285,093 g/mol
Schmelzpunkt: 110…112 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00573907
CAS-Nummer: 59609-59-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR