Methyl-4-(4-methoxyphenyl)-2,4-dioxobutanoate

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-(4-methoxyphenyl)-2,4-dioxobutanoate

OR14389-1G
APOSOR14389-1GEA 24.8 EUR
APOSOR14389-1G
Methyl-4-(4-methoxyphenyl)-2,4-dioxobutanoate
Methyl-4-(4-methoxyphenyl)-2,4-dioxobutanoate
Formel: C₁₂H₁₂O₅
Molecular Weight: 236,23 g/mol
Schmelzpunkt: 96…98 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04967380
CAS-Nummer: 39757-31-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR