3-(3-Oxo-1,3,4,5,6,7-hexahydro-2H-indazol-2-yl)benzenecarboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(1,3,4,5,6,7-Hexahydro-3-oxo-2H-indazol-2-yl)benzoic acid
OR14390-1G
OR14390-500MG
APOSOR14390-1GEA
36.4
EUR
APOSOR14390-1G
APOSOR14390-500MG
3-(3-Oxo-1,3,4,5,6,7-hexahydro-2H-indazol-2-yl)benzenecarboxylic acid
3-(3-Oxo-1,3,4,5,6,7-hexahydro-2H-indazol-2-yl)benzenecarboxylic acid
Formel:
C₁₄H₁₄N₂O₃ Molecular Weight: 258.28 g/mol Schmelzpunkt: 281 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659578 CAS-Nummer: 885949-86-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...