Methyl-5-iodsalicylat
Lieferant: Apollo Scientific
Synonyme:
Methyl 2-hydroxy-5-iodobenzoate
OR14411-25G
APOSOR14411-25GEA
74.1
EUR
APOSOR14411-25G
Methyl-5-iodsalicylat
Methyl-5-iodsalicylat
Formel:
C₈H₇IO₃ Molecular Weight: 278,05 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016462 CAS-Nummer: 4068-75-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...