2-Chloro-N-(4-methoxybenzyl)acetamide
Lieferant: Apollo Scientific
|
Achtung
|
OR14429-1G
APOSOR14429-1GEA
40.6
EUR
APOSOR14429-1G
2-Chloro-N-(4-methoxybenzyl)acetamide
2-Chloro-N-(4-methoxybenzyl)acetamide
Formel:
C₁₀H₁₂ClNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD02668858 CAS-Nummer: 81494-05-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...