1-(3-Methoxy-4-nitrophenyl)-1-ethanone

Lieferant: Apollo Scientific
Achtung

Synonyme: 3'-Methoxy-4'-nitroacetophenone

OR14436-1G OR14436-250MG
APOSOR14436-1GEA 97.3 EUR
APOSOR14436-1G APOSOR14436-250MG
1-(3-Methoxy-4-nitrophenyl)-1-ethanone
1-(3-Methoxy-4-nitrophenyl)-1-ethanone
Formel: C₉H₉NO₄
Molecular Weight: 195.18 g/mol
Schmelzpunkt: 71…73 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00075788
CAS-Nummer: 22106-39-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR