Methyl-3,5-dibromo-2-methoxybenzenecarboxylate

Lieferant: Apollo Scientific
Achtung

OR14437-1G
APOSOR14437-1GEA 81.2 EUR
APOSOR14437-1G
Methyl-3,5-dibromo-2-methoxybenzenecarboxylate
Methyl-3,5-dibromo-2-methoxybenzenecarboxylate
Formel: C₉H₈Br₂O₃
Molecular Weight: 323,98 g/mol
Schmelzpunkt: 52…54 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00189410
CAS-Nummer: 15790-59-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR