Ethylimidazo[1,2-a]pyridin-2-carboxylat 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR1446-5G
APOSOR1446-5GEA
72.5
EUR
APOSOR1446-5G
Ethylimidazo[1,2-a]pyridin-2-carboxylat 97%
Ethylimidazo[1,2-a]pyridin-2-carboxylat
Formel:
C₁₀H₁₀N₂O₂ Schmelzpunkt: 79…87 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02325394 CAS-Nummer: 38922-77-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...