Ethyl imidazo[2,1-b]thiazole-6-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR1447-1G
APOSOR1447-1GEA
0
EUR
APOSOR1447-1G
Ethyl imidazo[2,1-b]thiazole-6-carboxylate
Ethyl imidazo[2,1-b]thiazole-6-carboxylate
Formel:
C₈H₈N₂O₂S Schmelzpunkt: 87…91 °C Lagertemperatur: Kühlschrank |
CAS-Nummer:
64951-04-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...