6-(Hydroxymethyl)-2-methylquinoline

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Methylquinoline-6-methanol

OR14489-1G OR14489-5G
APOSOR14489-1GEA 192 EUR
APOSOR14489-1G APOSOR14489-5G
6-(Hydroxymethyl)-2-methylquinoline
6-(Hydroxymethyl)-2-methylquinoline
Formel: C₁₁H₁₁NO
Molecular Weight: 173.215 g/mol
Schmelzpunkt: 136…139 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659066
CAS-Nummer: 108166-02-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR