6-(Hydroxymethyl)-2-methylquinoline
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Methylquinoline-6-methanol
OR14489-1G
OR14489-5G
APOSOR14489-1GEA
192
EUR
APOSOR14489-1G
APOSOR14489-5G
6-(Hydroxymethyl)-2-methylquinoline
6-(Hydroxymethyl)-2-methylquinoline
Formel:
C₁₁H₁₁NO Molecular Weight: 173.215 g/mol Schmelzpunkt: 136…139 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659066 CAS-Nummer: 108166-02-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...