Ethyl 4-(5-methyl-1,2,4-oxadiazol-3-yl)benzoate

Lieferant: Apollo Scientific

OR1449-1G
APOSOR1449-1GEA 0 EUR
APOSOR1449-1G
Ethyl 4-(5-methyl-1,2,4-oxadiazol-3-yl)benzoate
Ethyl 4-(5-methyl-1,2,4-oxadiazol-3-yl)benzoate
Formel: C₁₂H₁₂N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD07368532
CAS-Nummer: 850375-01-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR