1-Boc-4-(4-formylphenyl)piperazine

Lieferant: Apollo Scientific

Synonyme: 1-Boc-4-(4-Formylphenyl)piperazine, 4-Piperazin-1-ylbenzaldehyde, N4-BOC protected

OR14490-25G
APOSOR14490-25GEA 177 EUR
APOSOR14490-25G
1-Boc-4-(4-formylphenyl)piperazine
1-Boc-4-(4-formylphenyl)piperazine
Formel: C₁₆H₂₂N₂O₃
Schmelzpunkt: 117…119 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05864663
CAS-Nummer: 197638-83-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR