Ethyl 5-(fur-2-yl)-1H-pyrazole-3-carboxylate

Lieferant: Apollo Scientific
Achtung

OR1451-1G
APOSOR1451-1GEA 0 EUR
APOSOR1451-1G
Ethyl 5-(fur-2-yl)-1H-pyrazole-3-carboxylate
Ethyl 5-(fur-2-yl)-1H-pyrazole-3-carboxylate
Formel: C₁₀H₁₀N₂O₃
Molecular Weight: 206.198 g/mol
Schmelzpunkt: 137…138 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06797478
CAS-Nummer: 34020-22-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR