3-(Bromomethyl)benzaldehyde 97%

Lieferant: Apollo Scientific
Gefahr

OR1453-1G OR1453-250MG OR1453-5G
APOSOR1453-1GEA 98.6 EUR
APOSOR1453-1G APOSOR1453-250MG APOSOR1453-5G
3-(Bromomethyl)benzaldehyde 97%
3-(Bromomethyl)benzaldehyde
Formel: C₈H₇BrO
Molecular Weight: 199.05 g/mol
Schmelzpunkt: 49…51 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06797497
CAS-Nummer: 82072-23-9
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR