3-(Bromomethyl)benzaldehyde 97%
Lieferant: Apollo Scientific
|
Gefahr
|
OR1453-1G
OR1453-250MG
OR1453-5G
APOSOR1453-1GEA
0
EUR
APOSOR1453-1G
APOSOR1453-250MG
APOSOR1453-5G
3-(Bromomethyl)benzaldehyde 97%
3-(Bromomethyl)benzaldehyde
Formel:
C₈H₇BrO Schmelzpunkt: 49…51 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797497 CAS-Nummer: 82072-23-9 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...