Fmoc-2-chloro-D-phenylalanine 95+%

Lieferant: Apollo Scientific

Synonyme: Fmoc-D-Phe(2-Cl)-OH

OR14543-5G
APOSOR14543-5GEA 126 EUR
APOSOR14543-5G
Fmoc-2-chloro-D-phenylalanine 95+%
Fmoc-2-chloro-D-phenylalanine
Formel: C₂₄H₂₀ClNO₄
Molecular Weight: 421,88 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00672547
CAS-Nummer: 205526-22-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR