Boc-3-chloro-L-phenylalanine 97%

Lieferant: Apollo Scientific

Synonyme: 3-Chloro-L-phenylalanine, N-BOC protected, Boc-Phe(3-Cl)-OH

OR14544-5G
APOSOR14544-5GEA 68.3 EUR
APOSOR14544-5G
Boc-3-chloro-L-phenylalanine 97%
Boc-3-chloro-L-phenylalanine
Formel: C₁₄H₁₈ClNO₄
Molecular Weight: 299,76 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00672515
CAS-Nummer: 114873-03-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR