Fmoc-4-chloro-L-phenylalanine 97%
Lieferant: Apollo Scientific
Synonyme:
Fmoc-4-chloro-Phe-OH, N-Fmoc-4-chloro-L-phenylalanine, Fmoc-Phe(4-Cl)-OH
OR14550-5G
APOSOR14550-5GEA
71.2
EUR
APOSOR14550-5G
Fmoc-4-chloro-L-phenylalanine 97%
Fmoc-4-chloro-L-phenylalanine
Formel:
C₂₄H₂₀ClNO₄ Molecular Weight: 421,877 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00080281 CAS-Nummer: 175453-08-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...