(S)-N-Boc-4-bromophenylalanine 97%
Lieferant: Apollo Scientific
Synonyme:
Boc-p-bromo-Phe-OH, Boc-Phe(4-Br)-OH
OR14556-25G
APOSOR14556-25GEA
133
EUR
APOSOR14556-25G
(S)-N-Boc-4-bromophenylalanine 97%
(S)-N-Boc-4-bromophenylalanine
Formel:
C₁₄H₁₈BrNO₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00237571 CAS-Nummer: 62129-39-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...