4,5-Methylenedioxy-2-nitrocinnamic acid

Lieferant: Apollo Scientific
Achtung

OR1456-25G OR1456-5G
APOSOR1456-25GEA 734 EUR
APOSOR1456-25G APOSOR1456-5G
4,5-Methylenedioxy-2-nitrocinnamic acid
4,5-Methylenedioxy-2-nitrocinnamic acid
Formel: C₁₀H₇NO₆
Schmelzpunkt: 280 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00063250
CAS-Nummer: 6315-90-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR