(2S)-2-({[(2-Methyl-2-propanyl)oxy]carbonyl}amino)-4-phenylbutansäure 97%

Lieferant: Apollo Scientific

Synonyme: N-Boc-L-homophenylalanine, (S)-2-(tert-Butoxycarbonylamino)-4-phenylbutyric Acid, Boc-Homophe-OH, (S)-2-(Boc-amino)-4-phenylbutyric Acid

OR14560-25G
APOSOR14560-25GEA 71.2 EUR
APOSOR14560-25G
(2S)-2-({[(2-Methyl-2-propanyl)oxy]carbonyl}amino)-4-phenylbutansäure 97%
(S)-2-((tert-Butoxycarbonyl)amino)-4-phenylbutanoic acid
Formel: C₁₅H₂₁NO₄
Molecular Weight: 279.34 g/mol
Schmelzpunkt: 76…80 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00076904
CAS-Nummer: 100564-78-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR