Boc-2-methyl-D-phenylalanine 95+%

Lieferant: Apollo Scientific

Synonyme: Boc-D-Phe(2-Me)-OH

OR14565-5G
APOSOR14565-5GEA 56.7 EUR
APOSOR14565-5G
Boc-2-methyl-D-phenylalanine 95+%
Boc-2-methyl-D-phenylalanine
Formel: C₁₅H₂₁NO₄
Molecular Weight: 279.34 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00671403
CAS-Nummer: 80102-29-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR