Fmoc-2-methyl-L-phenylalanine 95+%

Lieferant: Apollo Scientific

Synonyme: 2-Methyl-L-phenylalanine, N-FMOC protected, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(o-tolyl)propanoic acid

OR14566-5G
APOSOR14566-5GEA 230 EUR
APOSOR14566-5G
Fmoc-2-methyl-L-phenylalanine 95+%
Fmoc-2-methyl-L-phenylalanine
Formel: C₂₅H₂₃NO₄
Molecular Weight: 401.47 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00671391
CAS-Nummer: 211637-75-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR