N-Boc-3-methyl-L-phenylalanine

Lieferant: Apollo Scientific

Synonyme: Boc-Phe(3-Me)-OH

OR14568-5G
APOSOR14568-5GEA 115 EUR
APOSOR14568-5G
N-Boc-3-methyl-L-phenylalanine
N-Boc-3-methyl-L-phenylalanine
Formel: C₁₅H₂₁NO₄
Molecular Weight: 279.336 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01862944
CAS-Nummer: 114873-06-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR