3-Formyl-6-(methoxycarbonyl)-1H-indole, N-BOC protected

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-tert-Butyl 6-methyl-3-formyl-1H-indole-1,6-dicarboxylate

OR1457-1G
APOSOR1457-1GEA 140 EUR
APOSOR1457-1G
3-Formyl-6-(methoxycarbonyl)-1H-indole, N-BOC protected
3-Formyl-6-(methoxycarbonyl)-1H-indole, N-BOC protected
Formel: C₁₆H₁₇NO₅
Schmelzpunkt: 166…168 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD06797500
CAS-Nummer: 850374-95-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR